Skip to Content
Merck
All Photos(1)

Key Documents

PTCO050

Supelco

Pt-Co/Hazen/APHA Colour Reference Standard

ASTM D 1209 (ASTM colour 50)

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€336.00

€336.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€336.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€336.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is an analytical standard, manufactured and tested according to the ISO 9001 Quality system. Please note that the value stated on this page is nominal. All certified values for the current lots of all Pt-Co/Hazen/APHA colour reference standards can be found on the Paragon Scientific webpage.

Application

Pt-Co/Hazen/APHA (ASTM colour 50) colour reference standard is used for visual measurement of colour of solutions in the platinum-cobalt scale. It may be used as a colour standard for visual colour analysis by ASTM D 1209 test method.

Features and Benefits

  • Qualified under ISO 9001 Quality management system
  • Available with a certificate of analysis and MSDS
  • Full traceability to international standards with maximum data accuracy
  • Colour values compliant to ASTM D 1209 colour range
  • Supplied in a 500-mL pack size with tamper-evident packaging
  • 12-months of shelf life

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service