Skip to Content
Merck
All Photos(1)

Key Documents

PS737

Oxamyl

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13N3O3S
CAS Number:
Molecular Weight:
219.26
Beilstein:
2212753
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-737

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)O\N=C(/SC)C(=O)N(C)C

InChI

1S/C7H13N3O3S/c1-8-7(12)13-9-5(14-4)6(11)10(2)3/h1-4H3,(H,8,12)/b9-5-

InChI key

KZAUOCCYDRDERY-UITAMQMPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Strathmann et al.
Environmental science & technology, 35(12), 2461-2469 (2001-07-04)
The degradation of two oxime carbamate pesticides, oxamyl and methomyl, was investigated in anoxic solutions containing various metal ions and reducing agents. In reagent-free solutions, these carbamates degrade slowly via base-catalyzed elimination. Rates of carbamate degradation are accelerated by Fe(II)
Rachel K Osborn et al.
Pest management science, 66(3), 253-261 (2009-10-29)
The potential for enhanced degradation of the carbamoyloxime nematicides aldicarb and oxamyl and the organophosphate fosthiazate was investigated in 35 UK agricultural soils. Under laboratory conditions, soil samples received three successive applications of nematicide at 25 day intervals. The second
Gerardo A Anguiano et al.
Environmental toxicology, 25(4), 327-332 (2009-05-19)
Acetylcholinesterase (AChE) activity has been used to test the exposure of mollusk bivalves to pesticides and other pollutants. The Pacific oyster Crassostrea gigas is a species with a worldwide distribution, and it has a high commercial value. The use of
F Lamberti et al.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen), 67(3), 699-702 (2003-04-17)
Bunches of 25 g of tomato galled roots containing ca. 270 eggs and juveniles/g of Meloidogyne incognita were buried on 29 September 2001 in gauze bags at the depth of 15, 30 and 45 cm in a sandy loam. The
Connie Hiers et al.
The Journal of the Arkansas Medical Society, 106(7), 134-136 (2009-12-18)
This is a case report and literature review of a 30.8 pound cystosarcoma phyllodes tumor resected in a 54-year-old female. Starting at age 19, the patient had noted a small breast tumor that continued to grow over the years. Pathology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service