Skip to Content
Merck
All Photos(1)

Key Documents

PS690-250MG

Hexachlorobenzene

analytical standard

Synonym(s):

HCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl6
CAS Number:
Molecular Weight:
284.78
Beilstein:
1912585
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-690

bp

323-326 °C (lit.)

mp

227-229 °C (lit.)

SMILES string

Clc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CKAPSXZOOQJIBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manas K Panda et al.
Nature chemistry, 7(1), 65-72 (2014-12-18)
The exceptional mechanical flexibility observed with certain organic crystals defies the common perception of single crystals as brittle objects. Here, we describe the morphostructural consequences of plastic deformation in crystals of hexachlorobenzene that can be bent mechanically at multiple locations
Noël J Diepens et al.
Environmental science & technology, 49(22), 13586-13595 (2015-10-16)
Whole sediment toxicity tests play an important role in environmental risk assessment of organic chemicals. It is not clear, however, to what extent changing microbial community composition and associated functions affect sediment test results. We assessed the development of bacterial
Lesa L Aylward et al.
Regulatory toxicology and pharmacology : RTP, 58(1), 25-32 (2010-06-16)
Recent efforts worldwide have resulted in a growing database of measured concentrations of chemicals in blood and urine samples taken from the general population. However, few tools exist to assist in the interpretation of the measured values in a health
Jonathan L Barber et al.
The Science of the total environment, 349(1-3), 1-44 (2005-07-12)
Hexachlorobenzene (HCB) is considered here as a 'model persistent organic pollutant.' Data on its sources, emissions, environmental levels and distributions and trends are compiled and used to assess its fate and behaviour in the global environment. Consideration is given as
C C Michielsen et al.
Environmental health perspectives, 107 Suppl 5, 783-792 (1999-09-30)
Hexachlorobenzene (HCB) is a persistent environmental pollutant. The toxicity of HCB has been extensively studied after an accidental human poisoning in Turkey and more recently it has been shown that HCB has immunotoxic properties in laboratory animals and probably also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service