Skip to Content
Merck
All Photos(1)

Key Documents

PS610

Paraoxon

analytical standard

Synonym(s):

O,O-Diethyl O-(4-nitrophenyl) phosphate, Diethyl p-nitrophenyl phosphate, Paraoxon, Paraoxon-ethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(OC2H5)2
CAS Number:
Molecular Weight:
275.20
Beilstein:
1915526
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-610

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.51 (lit.)

density

1.274 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCOP(=O)(OCC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C10H14NO6P/c1-3-15-18(14,16-4-2)17-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3

InChI key

WYMSBXTXOHUIGT-UHFFFAOYSA-N

Gene Information

human ... PON1(5444)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Potent irreversible acetylcholinesterase inhibitor

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. Is Product PS610, Paraoxon - analytical standard, a solid or a liquid?

    This is an oil or liquid at room temperature.

  5. What is Product PS610, Paraoxon - analytical standard, soluble in?

    This is freely soluble in diethyl ether, and in most other organic solvents.

  6. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  7. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Zoran Radić et al.
The Biochemical journal, 450(1), 231-242 (2012-12-12)
In the present paper we show a comprehensive in vitro, ex vivo and in vivo study on hydrolytic detoxification of nerve agent and pesticide OPs (organophosphates) catalysed by purified hBChE (human butyrylcholinesterase) in combination with novel non-pyridinium oxime reactivators. We
Krystle L Yozzo et al.
Aquatic toxicology (Amsterdam, Netherlands), 126, 346-354 (2012-10-11)
Using paraoxon as a reference acetylcholinesterase (AChE) inhibitor, the objective of this study was to develop an adverse outcome pathway (AOP) that provided quantitative linkages across levels of biological organization during zebrafish embryogenesis. Within normal zebrafish embryos, we first demonstrated
Francesca Cavalieri et al.
ACS applied materials & interfaces, 5(2), 464-471 (2012-12-26)
Air-filled lysozyme microbubbles (LSMBs) were engineered as a support for the immobilization of gold nanoparticles and an enzyme, alkaline phosphatase, in order to develop micro-antimicrobial and biosensing devices. Gold nanoparticles immobilized on LSMBs significantly improved the antimicrobial efficacy of the
Guanglei Fu et al.
Talanta, 103, 110-115 (2012-12-04)
Highly sensitive colorimetric detection of organophosphate pesticides (OPs) was developed using Cu (I)-catalyzed click chemistry as the colorimetric signal amplification process between the acetylcholine esterase-acetylthiocholine system (AChE-ATCl) and azide- terminal alkyne-functionalized Au NPs as the colorimetric probe. It was demonstrated
D E Lorke et al.
Journal of applied toxicology : JAT, 29(6), 459-469 (2009-07-16)
K-oximes have recently been developed in the search for efficacious broad-band reactivators of acetylcholinesterase (AChE) inhibited by organophosphorus compounds (OPC). Before clinical use, their toxicity and efficacy need to be assessed, and there is clear demand for simple in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service