Skip to Content
Merck
All Photos(1)

Key Documents

PS2044-250MG

Thidiauron

Synonym(s):

Thidiazuron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N4OS
CAS Number:
Molecular Weight:
220.25
Beilstein:
1078092
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-2044

SMILES string

O=C(Nc1ccccc1)Nc2cnns2

InChI

1S/C9H8N4OS/c14-9(12-8-6-10-13-15-8)11-7-4-2-1-3-5-7/h1-6H,(H2,11,12,14)

InChI key

HFCYZXMHUIHAQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Cutri et al.
Mechanisms of development, 130(1), 61-69 (2012-06-05)
Tendrils can be found in different plant species. In legumes such as pea, tendrils are modified leaves produced by the vegetative meristem but in the grape vine, a same meristem is used to either form a tendril or an inflorescence.
Dmytro P Yevtushenko et al.
Plant cell reports, 29(3), 211-221 (2010-01-21)
Many economically important species of Populus, especially those in sections Aigeiros and Tacamahaca, remain recalcitrant to genetic transformation. In this study, a simple and reliable protocol was developed for the efficient Agrobacterium-mediated transformation of a difficult-to-transform, but commercially viable, hybrid
Alexander V Babosha
Journal of plant physiology, 166(17), 1892-1903 (2009-07-14)
Dose-response relationship between resistance of wheat seedlings (Triticum aestivum, cultivar Zarya) to Erysiphe graminis f. sp. tritici Marchal. (Syn. Blumeria graminis), a causal organism of wheat powdery mildew and exogenous zeatin has been investigated. Two-week-old seedlings were inoculated with the
Hemaid Ibrahim Soliman et al.
GM crops, 1(1), 40-51 (2010-01-01)
Fig is one of the most important fruit trees in Egypt. It used to constitute the major source of income for the inhabitants of the western north coast of Egypt. Since 1993 fig cultivations were threatened by a number of
Sureerat Lo-apirukkul et al.
Journal of natural medicines, 66(2), 265-270 (2011-08-19)
We studied the effects of explant types, plant growth regulators, and sucrose concentrations on shoot and microrhizome inductions of Curcuma comosa Roxb., an important Thai medicinal plant for women's health. Explant types significantly affected shoot induction of the plant. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service