Skip to Content
Merck
All Photos(1)

Key Documents

PS1024

Profenofos

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15BrClO3PS
CAS Number:
Molecular Weight:
373.63
Beilstein:
2150258
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-1024

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCSP(=O)(OCC)Oc1ccc(Br)cc1Cl

InChI

1S/C11H15BrClO3PS/c1-3-7-18-17(14,15-4-2)16-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3

InChI key

QYMMJNLHFKGANY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Kavitha et al.
Ecotoxicology and environmental safety, 72(6), 1727-1733 (2009-06-09)
The euryhaline fish, Oreochromis mossambicus was exposed to sub-lethal concentration (30 microg/L) of profenofos (PF) for 28 days and allowed to recover for 7 days. Responses were evaluated through antioxidant enzyme activities in various tissues of fish. Glutathione-S-transferase (GST) and
Kriengkrai Satapornvanit et al.
Chemosphere, 74(9), 1209-1215 (2008-12-24)
A bioassay was developed using post-larvae of freshwater prawn Macrobrachium rosenbergii (length 9-10 mm) in order to determine the toxicity of profenofos, dimethoate, chlorpyrifos, carbendazim and zinc. This was the first study in the tropics with M. rosenbergii, particularly at
Irani Mukherjee et al.
Bulletin of environmental contamination and toxicology, 88(3), 338-343 (2011-11-08)
Persistence behavior of two combination mix formulations of insecticides viz. profenofos + cypermethrin (Roket 44EC) and chlorpyrifos + cypermethrin (Nacraj-505) were studied in eggplant fruits following spray application at recommended and double dose. Half-life values were calculated from first order
Na Gao et al.
The Analyst, 137(5), 1252-1258 (2012-01-21)
A quartz crystal microbalance (QCM) sensor based on molecularly imprinted ultra-thin films was developed for detecting profenofos in real samples. Films prepared by physical entrapment (MIP-A) and in situ self-assembly (MIP-B) were compared. The results indicated that the best sensing
Susmita Ganguly et al.
Ecotoxicology (London, England), 19(1), 85-95 (2009-07-21)
In this study, attention was paid to investigate the effect of organophosphate insecticides, profenofos 40% EC, methyl parathion (metacid) 50% EC, and neem-based product nimbecidine 0.03% EC (from Azadirachta indica) on somatic chromosomal behavior, level of leaf protein, and activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service