Skip to Content
Merck
All Photos(1)

Key Documents

PHR9000

Supelco

2-[1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,4-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazol-3-ium-3-yl]-2-methylpropanoate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Miconazole N-(2-Methyl)propanoate, Miconazole Nitrate impurity E (PhEur)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€449.00

€449.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€449.00

About This Item

Empirical Formula (Hill Notation):
C22H20Cl4N2O3
Molecular Weight:
502.22
UNSPSC Code:
41116107
NACRES:
NA.24

€449.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical small molecule

SMILES string

ClC1=CC(Cl)=CC=C1COC(CN2C=[N+](C(C)(C)C([O-])=O)C=C2)C3=C(Cl)C=C(Cl)C=C3

InChI

1S/C22H20Cl4N2O3/c1-22(2,21(29)30)28-8-7-27(13-28)11-20(17-6-5-16(24)10-19(17)26)31-12-14-3-4-15(23)9-18(14)25/h3-10,13,20H,11-12H2,1-2H3

InChI key

XPEIFDJIVDKSMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

These Reference Materials are suitable for use in several analytical applications including, but not limited to, method development for qualitative and quantitative analyses, daily calibration, and routine quality control testing.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service