Skip to Content
Merck
All Photos(1)

Documents

PHL89498

trans-p-Coumaric acid

phyproof® Reference Substance

Synonym(s):

p-Coumaric acid, trans-4-Hydroxycinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16
Beilstein:
2207383
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

mp

214 °C (dec.) (lit.)

SMILES string

OC(=O)\C=C\c1ccc(O)cc1

InChI

1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)/b6-3+

InChI key

NGSWKAQJJWESNS-ZZXKWVIFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Hydroxycinnamic acid found in many fruits and vegetables.

Other Notes

This compound is commonly found in plants of the genus: curcuma mentha zingiber

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service