Skip to Content
Merck
All Photos(1)

Documents

PHL80496

gamma-Mangostin

phyproof® Reference Substance

Synonym(s):

1,3,6,7-Tetrahydroxy-2,8-bis(3,3-dimethylallyl)xanthone, Demethylmangostin, Normangostin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O6
CAS Number:
Molecular Weight:
396.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

OC1=CC(OC2=CC(O)=C(CC=C(C)C)C(O)=C2C3=O)=C3C(CC=C(C)C)=C1O

InChI

1S/C23H24O6/c1-11(2)5-7-13-15(24)9-18-20(22(13)27)23(28)19-14(8-6-12(3)4)21(26)16(25)10-17(19)29-18/h5-6,9-10,24-27H,7-8H2,1-4H3

InChI key

VEZXFTKZUMARDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Xanthone derivative isolated from the mangosteen hull (Garcinia mangostana). Exhibits antioxidant and anticancer properties. Mangosteen xanthones inhibit inflammation in human adipocytes which is important in understanding obesity-associated inflammation and the development of insulin resistance.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service