Skip to Content
Merck
All Photos(1)

Key Documents

G006119

HPLC Analysis of Bovine Serum Albumin (BSA) on Zenix® SEC-300 versus SRT® SEC-300, Effect of Particle Size on Resolution

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: SRT SEC-300, 30 cm x 7.8 mm I.D., 5 μm, 300 Å particles (Z777051)
column: Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm, 300 Å particles (Z777033)
mobile phase: 150 mM sodium phosphate, pH 7
flow rate: 1 mL/min
column temp.: ambient
detector: UV, 214 nm
injection: 10 μL
sample: 5 mg/mL, BSA

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

SRT is a registered trademark of Sepax Technologies
Zenix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service