Skip to Content
Merck
All Photos(1)

Key Documents

G005833

HPLC Analysis of Spice Cannabinoids JWH-018, JWH-073 and Metabolites on Ascentis® Express C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 μm particles (53814-U)
mobile phase: [A] 0.1% formic acid in water; [B] 0.1% formic acid in water:acetonitrile (10:90)
gradient: 20% B held for 3 min; 20 to 100% B in 0.1 min; 100% B held for 2 min
flow rate: 0.5 mL/min
pressure: 2150 psi (148 bar)
column temp.: 35 °C
detector: UV, 315 nm
injection: 2 μL
sample: 17 μg/mL each in acetonitrile

suitability

application for HPLC

application(s)

forensics and toxicology

Analysis Note

Synthetic cannabinoids (e.g. "Spice") are a type of designer drug that provides a cannabis-type high. New synthetic cannabinoids are continually being introduced as suppliers tweak the molecular structures. The ability to rapidly and reliably identify the continually changing population of these compounds is a significant analytical challenge facing forensic chemists. A rapid separation of six of these compounds on an Ascentis Express C18 column is shown here. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    JWH-073 4-Butanoic acid metabolite solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    JWH-018 5-Pentanoic acid metabolite solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    JWH-073 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
    JWH-018 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service