Skip to Content
Merck
All Photos(1)

Documents

G004756

GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 85 μm Polyacrylate Fiber, Fast GC Analysis

suitable for GC, application for SPME

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable
solid phase microextraction (SPME): suitable

test parameters

sample/matrix: 500 ug/L of each compound in 2 mL 0.05M phosphate buffer, pH=11, plus sodium chloride (NaCl) to 25% (w/v)
SPME fiber: 85 μm Polyacrylate (57304)
extraction: immersion, 450 rpm stirring at room temp. for 15 min
desorption process: 250 °C for 3 min
column: SLB-5ms, 20 m x 0.18 mm I.D. x 0.36 μm (28576-U)
oven: 100 °C (1 min), 20 °C/min to 325 °C (5 min)
MSD interface: 325 °C
carrier gas: helium, 1.5 mL/min. constant
liner: 0.75 mm I.D. SPME

suitability

application for SPME

application(s)

forensics and toxicology

Legal Information

SLB is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service