Skip to Content
Merck
All Photos(1)

Documents

G004737

HPLC Analysis of Triazole Antifungal Agent Enantiomers on Astec® CYCLOBOND I 2000 HP-RSP

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Astec CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase: [A] 0.1% formic acid; [B] acetonitrile; (70:30, A:B)
flow rate: 1 mL/min
detector: UV, 230 nm
injection: 1 μL
sample: 0.1 mg/mL in methanol
peaks 1&2: enilconazole
peaks 2&3: eberconazole
peaks 4&5: miconazole
peaks 6&7: ketoconazole

suitability

application for HPLC

application(s)

chemicals and industrial polymers

Analysis Note

Note: Peak 2 of enilconzaole coelutes with peak 1 of eberconazole under these conditions.

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service