Skip to Content
Merck
All Photos(1)

Key Documents

CRM68466

Supelco

Fluazifop-butyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€84.70

€84.70


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
€84.70

About This Item

Empirical Formula (Hill Notation):
C19H20F3NO4
CAS Number:
Molecular Weight:
383.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€84.70


Please contact Customer Service for Availability

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

quality

Racemate

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

-10 to -25°C

SMILES string

CCCCOC(=O)C(C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1

InChI

1S/C19H20F3NO4/c1-3-4-11-25-18(24)13(2)26-15-6-8-16(9-7-15)27-17-10-5-14(12-23-17)19(20,21)22/h5-10,12-13H,3-4,11H2,1-2H3

InChI key

VAIZTNZGPYBOGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Fluazifop-butyl belongs to the class of aryloxyphenoxy-propionate (AOPP) herbicides that is used for postemergence control of annual and perennial grasses in a range of broad-leaved crops. When applied, this selective herbicide is absorbed by leaves and acts by inhibiting the acetyl-CoA carboxylase (ACCase) enzyme activity in plastids. Fluazifop-butyl rapidly degrades to the free acid form –fluazifop, a major chiral metabolite.

Fluazifop is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. The maximum residue level (MRLs) for Fluazifop-P (sum of all the constituent isomers of fluazifop, its esters, and conjugates, expressed as fluazifop) has been set from 0.01 mg/kg to 9 mg/kg for products of plants and animal origin following Regulation (EU) 2018/1514.

Application

Fluazifop-butyl may be used as reference material for the following:
  • Separation of the enantiomers of fluazifop-butyl and fluazifop by LC/MS/MS, followed by investigating the stereoselective degradation and transformation of the enantiomers of the herbicide fluazifop-butyl in soil and water
  • To investigate the effects of herbicide fluazifop-butyl on broad-leaved weed, Acanthospermum hispidum, and study the mechanism of action of the herbicide on the weed
  • To develop a residue analysis method for the determination of fluazifop-butyl and its chiral metabolite fluazifop in soil and vegetable and to investigate the enantioselective kinetics of fluazifop-butyl and the optical pure fluazifop-P-butyl
  • To determine the mechanism of fluazifop-butyl action on bristly starbur and investigate the ethylene production and its involvement in lipid peroxidation of the membrane
  • Simultaneous determination of fluazifop-butyl and fluazifop in soil samples using HPLC

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service