Skip to Content
Merck
All Photos(1)

Documents

Celite® Analytical Filter Aid II (CAFA II) Inorganic Adsorbent

SLE ion exchange

Synonym(s):

Diatomaceous earth, calcined, CAFA II 100g (Celite® Analytical Filter Aid II)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
40161804
PubChem Substance ID:

product name

Celite® Analytical Filter Aid II (CAFA II),

packaging

pkg of 1 × 1 kg (11486-U)
pkg of 1 × 100 g (11484-U)
pkg of 1 × 500 g (11485-U)

technique(s)

LPLC: suitable

matrix

Silica

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CAFA II, a high-purity Celite® analytical filter aid, is a replacement for the original CAFA material. It is of higher purity and consistency as compared to the original CAFA. But, its filtration and physical characteristics are nearly indistinguishable from CAFA. It has a filtration efficiency in the range of 0.5-1.0 microns.

Application

Celite® analytical aid II (CAFA II) find its use as a high-purity filter-aid in pharmaceutical applications. It may be used to filter reaction mixtures in the interesterification of fat blends to study their physical properties.

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service