Skip to Content
Merck
All Photos(1)

Documents

BP889

N-Demethylerythromycin A

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

DEMA, N-Demethylerythromycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H65NO13
CAS Number:
Molecular Weight:
719.90
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

erythromycin

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

−20°C

InChI

1S/C36H65NO13/c1-13-24-36(10,44)29(40)19(4)26(38)17(2)15-34(8,43)31(50-33-27(39)23(37-11)14-18(3)46-33)20(5)28(21(6)32(42)48-24)49-25-16-35(9,45-12)30(41)22(7)47-25/h17-25,27-31,33,37,39-41,43-44H,13-16H2,1-12H3

InChI key

TXOOBKBDBNRQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

N-Demethylerythromycin A BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service