Skip to Content
Merck
All Photos(1)

Key Documents

97610

Supelco

1,3-Dioxolane

Synonym(s):

Ethylene glycol methylene ether, Formaldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€60.30

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€60.30

About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Beilstein:
102453
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

2.6 (vs air)

vapor pressure

70 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

525 °F

technique(s)

HPLC: suitable

refractive index

n20/D 1.401 (lit.)

bp

75-76 °C/1.013 hPa

mp

−95 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

C1COCO1

InChI

1S/C3H6O2/c1-2-5-3-4-1/h1-3H2

InChI key

WNXJIVFYUVYPPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service