Skip to Content
Merck
All Photos(1)

Key Documents

96008

Supelco

Cyclohexyl methacrylate

Synonym(s):

Methacrylic acid cyclohexyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€60.30

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€60.30

About This Item

Linear Formula:
CH2=C(CH3)COOC6H11
CAS Number:
Molecular Weight:
168.23
Beilstein:
2045235
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.21

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

contains

10-80 ppm monomethyl ether hydroquinone as inhibitor as stabilizer

technique(s)

HPLC: suitable

refractive index

n20/D 1.458 (lit.)

bp

68-70 °C/4 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(=C)C(=O)OC1CCCCC1

InChI

1S/C10H16O2/c1-8(2)10(11)12-9-6-4-3-5-7-9/h9H,1,3-7H2,2H3

InChI key

OIWOHHBRDFKZNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service