Skip to Content
Merck
All Photos(1)

Key Documents

95323

Supelco

1-Amino-3-(aminomethyl)-3,5,5-trimethylcyclohexane

analytical standard

Synonym(s):

5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans, Isophorone diamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€60.30

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€60.30

About This Item

Linear Formula:
H2NC6H7(CH3)3CH2NH2
CAS Number:
Molecular Weight:
170.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

98.0% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.488 (lit.)

bp

247 °C (lit.)

mp

10 °C (lit.)

density

0.922 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC1(C)CC(N)CC(C)(CN)C1

InChI

1S/C10H22N2/c1-9(2)4-8(12)5-10(3,6-9)7-11/h8H,4-7,11-12H2,1-3H3

InChI key

RNLHGQLZWXBQNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service