Skip to Content
Merck
All Photos(1)

Key Documents

94708

Supelco

1,5-Dihydroxynaphthalene

analytical standard

Synonym(s):

1,5-Naphthalenediol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€60.30

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€60.30

About This Item

Linear Formula:
C10H6(OH)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
2044951
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

€60.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0 % (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

259-261 °C (dec.) (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Oc1cccc2c(O)cccc12

InChI

1S/C10H8O2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6,11-12H

InChI key

BOKGTLAJQHTOKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service