Skip to Content
Merck
All Photos(1)

Key Documents

90864

Supelco

Sodium stearate

analytical standard

Synonym(s):

Octadecanoic acid sodium salt, Stearic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COONa
CAS Number:
Molecular Weight:
306.46
Beilstein:
3576813
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98.0% (GC)

form

powder

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

[Na+].CCCCCCCCCCCCCCCCCC([O-])=O

InChI

1S/C18H36O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2-17H2,1H3,(H,19,20);/q;+1/p-1

InChI key

RYYKJJJTJZKILX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service