Skip to Content
Merck
All Photos(1)

Key Documents

90126R

Supelco

Aroclor 1242 solution

1000 μg/mL in hexane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

packaging

ampule of 1 mL

concentration

1000 μg/mL in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

room temp

InChI

1S/C12H6Cl4/c13-7-1-3-9(11(15)5-7)10-4-2-8(14)6-12(10)16/h1-6H

InChI key

QORAVNMWUNPXAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Tremaroli et al.
FEMS microbiology ecology, 74(2), 291-301 (2010-09-18)
Pseudomonas pseudoalcaligenes KF707 is a polychlorinated biphenyls (PCBs) degrader, also tolerant to several toxic metals and metalloids. The work presented here examines for the first time the chemotactic response of P. pseudoalcaligenes KF707 to biphenyl and intermediates of the PCB
M B Voltura et al.
Archives of environmental contamination and toxicology, 52(2), 264-269 (2006-12-13)
Studies of the impact of environmental contaminants on reproduction have typically focused on effects on fertility and subsequent reproductive failure. Contaminants may also impact reproductive output or other aspects of life history through effects on resource acquisition or allocation. We
Young-Cheol Cho et al.
Journal of microbiology (Seoul, Korea), 43(2), 166-171 (2005-05-10)
In order to determine the effects of sulfate concentration on the anaerobic dechlorination of polychlorinated biphenyls, sediments spiked with Aroclor 1242 were made into slurries using media which had various sulfate concentrations ranging from 3 to 23 mM. The time
Martha Gayosso-Canales et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 46(3), 298-305 (2011-02-11)
A 2(III)(7-3) fractional factorial experimental design was used to establish 16 culture media, with and without PCBs to enhance the activities of laccase (Lac), manganese peroxidase (MnP), and versatile peroxidase (VP) produced by the white rot fungus Pleurotus ostreatus. The
Lars Rehmann et al.
Biotechnology and bioengineering, 99(5), 1273-1280 (2007-10-12)
This article demonstrates the feasibility of a novel process concept for the remediation of PCB contaminated soil. The proposed process consists of PCB extraction from soil using solid polymer beads, followed by biodegradation of the extracted PCBs in a solid-liquid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service