Skip to Content
Merck
All Photos(1)

Key Documents

843271

TOYOPEARL® QAE-550C Bulk Media

phase ammonium ion, quaternary, bottle of 100 mL, 100 μm particle size

Synonym(s):

TOYOPEARL® QAE-550C Anion Exchange Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
Product 843271 is not currently sold in your country. Contact Technical Service

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 43271

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ammonium ion, quaternary phase

particle size

100 μm

operating pH

2-13

capacity

70 g/L±10 adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl QAE-550 resin is a strong anion exchanger useful for purification of proteins up to 100kD. It is used for high throughput capture of feedstocks.

Specifications

Clean in place with 0.1 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service