Skip to Content
Merck
All Photos(1)

Documents

823513

Supelco

TSKgel® FcR-IIIA-NPR (5 µm) HPLC Columns

L x I.D. 7.5 cm × 4.6 mm, Glycoprotein phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product name

TSKgel® FcR-IIIA-NPR HPLC Column, PEEK, L × I.D. 7.5 cm × 4.6 mm, 5 μm particle size

ligand

(modified recombinant FcyRIIIA)

material

PEEK

product line

TSKgel®

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 4.6 mm

matrix

polymethacrylate

matrix active group

Glycoprotein phase

particle size

5 μm

operating pH range

4.0-8.0

separation technique

affinity

storage temp.

2-8°C

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fast assessment of antibody-dependent cell-mediated cytotoxicity (ADCC) and glycoform pattern of a therapeutic antibody rituximab by FcγRIIIa affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service