Skip to Content
Merck
All Photos(1)

Key Documents

822832

TSKgel® Protein C4-300 (3 µm) HPLC Columns

L x I.D. 15 cm × 2 mm, C4 (butyl) phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€971.00

€971.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€971.00

About This Item

UNSPSC Code:
41115711

€971.00


Please contact Customer Service for Availability

Product Name

TSKgel® Protein C4-300 HPLC Column, phase C4 (butyl), L × I.D. 15 cm × 2 mm, 3 μm particle size

material

stainless steel

Agency

suitable for USP L26

product line

TSKgel®

manufacturer/tradename

Tosoh 22832

extent of labeling

3% Carbon, endcapped with trimethylsilane

parameter

no limitation salt concentration
0-100% organic solvent
1 mL/min standard flow rate
10-50 °C temperature
300 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 2 mm

surface area

100 m2/g

matrix

fully porous particle

matrix active group

C4 (butyl) phase

particle size

3 μm

operating pH

1.5-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Protein C4-300 columns are the most recent addition to the extensive line of Reversed Phase columns from Tosoh Bioscience. The high efficiency columns are designed for the optimal recovery and resolution of proteins such as recombinant proteins, antibody fragments or PEGylated proteins. The 300 Å pore size of the TSKgel Protein C4-300 columns are ideal for the separation of proteins. A particle size of 3 μm and optimized ligand density and alkyl length result in better protein and peptide resolution compared to other leading RP-C4 HPLC phases. Latest surface modification techniques and endcapping of residual silanol groups reduce undesirable secondary interactions and peak tailing.

Physical form

Shipped in acetonitrile.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service