Skip to Content
Merck
All Photos(1)

Key Documents

822819

TOYOPEARL® MX-Trp-650M Bulk Media

phase tryptophan, bottle of 1 L, 75 μm particle size

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€5,020.00

€5,020.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
€5,020.00

About This Item

UNSPSC Code:
41106500

€5,020.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 22819

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

tryptophan phase

particle size

75 μm

operating pH

3-13

capacity

75 g/L adsorption capacity (γ globulin)

separation technique

mixed mode

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl MX-Trp-650M is a high capacity mixed-mode resin used for the purification of monoclonal antibodies and other proteins. The multimodal resin maintains dynamic binding capacity (DBC) at elevated feedstock or buffer conductivities. Along with fast binding and elution kinetics, Toyopearl MX-Trp-650M minimizes the amount of WFI needed for either feedstock dilution, or column elution, leading to lower in-process pool volumes and higher throughput.
Toyopearl MX-Trp-650M maintains a high DBC even after extended CIP (clean in place) cycles using 0.5 mol/L NaOH.

Specifications

Clean in place with 0.5 M NaOH.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service