Skip to Content
Merck
All Photos(1)

Key Documents

822792

TSKgel® SuperOligoPW (3 µm) HPLC Columns

L x I.D. 15 cm × 6 mm, hydroxylated methacrylate phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,640.00

€2,640.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€2,640.00

About This Item

UNSPSC Code:
41115711

€2,640.00


Please contact Customer Service for Availability

Product Name

TSKgel® SuperOligoPW HPLC Column, phase hydroxylated methacrylate, L × I.D. 15 cm × 6 mm, 3 μm particle size

material

stainless steel

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 22792

parameter

≤0.5 M salt concentration
≤20% polar organic solvent
0.6 mL/min flow rate
10-80 °C temperature
50 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 6 mm

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylate phase

particle size

3 μm

operating pH

2-12

separation technique

size exclusion (SEC)

General description

The TSKgel® SuperOligoPW column is designed for the determination of molecular mass of aqueous oligomers, particularly oligosaccharides, and low molecular weight aqueous polymers. Compared to the TSKgel G-Oligo-PW column, the three micron particle size and semi-micro dimensions (15 cm x 6 mm ID) of the TSKgel SuperOligoPW column reduces analysis time by a factor of two while maintaining resolution and lowering solvent consumption.Since the packing material in the TSKgel SuperOligoPW columns is more hydrophilic compared with TSKgel G-Oligo-PW columns, an even wider range of water-soluble polymers can be eluted without the need to add organic solvent to the eluent.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service