Skip to Content
Merck
All Photos(1)

Key Documents

821875

TSKgel® G6000PWXL-CP (13 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,580.00

€2,580.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€2,580.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

€2,580.00


Please contact Customer Service for Availability

Product Name

TSKgel® G6000PWXL-CP HPLC Column, phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 13 μm particle size

material

stainless steel hardware

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 21875

parameter

≤0.5 M salt concentration
≤20% organic solvent
1 mL/min flow rate
20 bar max. pressure
60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

13 μm

pore size

>1000 Å pore diameter

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel PWXL-CP columns were specifically developed for the analysis of water-soluble cationic polymers. Composed of polymethacrylate beads, cationic groups are introduced on the surface of the TSKgel PWXL-CP packing material to prevent adsorption of cationic polymers and allow elution under low salt conditions. These columns show high theoretical plate numbers, linear calibration curves and high durability because the base resin is the same as that used in the TSKgel PWXL series columns. Three columns are available within the TSKgel PWXL-CP series, each with a different particle size, separation range and exclusion limit, allowing polymers within a wide molecular mass range to be separated and characterized.

Application

Exclusion limit (Da): 20,000,000
Separation range (Da) (PEO,PEG): 1,000~10,000,000

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Separation of various acids and compounds by molecular weight: PAA, PEI, PIDADMACl, PAS, cationic dextran, chitosan

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service