Skip to Content
Merck
All Photos(1)

Key Documents

821303

TOYOPEARL® PPG-600M Bulk Media

phase polypropylene glycol, bottle of 1 L, 65 μm particle size

Synonym(s):

TOYOPEARL® PPG-600 HIC Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€4,370.00

€4,370.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
€4,370.00

About This Item

UNSPSC Code:
41106500

€4,370.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 21303

parameter

≤50% organic solvent
3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

polypropylene glycol phase

particle size

65 μm

operating pH

1-13

capacity

55 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl PPG-600 (polypropylene glycol) resin has the second highest hydrophilicity of the HIC ligands offered by Tosoh Bioscience. It has been optimized for the purification of monoclonal antibodies and is used as an intermediate purification or polishing step. It has extremely good capacity and recovery for mAbs.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service