Skip to Content
Merck
All Photos(1)

Key Documents

82019-U

Hamilton® PRP-X200 (10 µm) HPLC Columns

L × I.D. 15 cm × 4.1 mm, stainless steel hardware

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€783.00

€783.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€783.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

€783.00


Please contact Customer Service for Availability

Product Name

Hamilton PRP-X200 HPLC Column, 10 μm particle size, L × I.D. 15 cm × 4.1 mm, stainless steel hardware

material

stainless steel column

Agency

suitable for USP L22

manufacturer/tradename

Hamilton

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.1 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

sulfonic acid phase

particle size

10 μm

pore size

100 Å

operating pH

1-14

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

The Hamilton PRP-X200, cation exchange columns are used in the separation of inorganic and organic, mono or divalent cations.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service