Skip to Content
Merck
All Photos(1)

Key Documents

819660

TSKgel® SuperHZM-N (3 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, polystyrene-divinylbenzene phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,330.00

€2,330.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€2,330.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

€2,330.00


Please contact Customer Service for Availability

Product Name

TSKgel® SuperHZM-N HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 3 μm particle size

material

stainless steel hardware

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 19660

parameter

0.4 mL/min flow rate
35 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

3 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SuperHZM-N mixed-bed columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the analysis of organic-soluble polymers with high molecular weights of less than 700,000 Da. The THF shipping solvent may be replaced by a limited number of organic solvents.

Physical form

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service