Skip to Content
Merck
All Photos(1)

Key Documents

819317

TSKgel® SuperAW4000 (6 µm) HPLC Columns

L x I.D. 15 cm × 6 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® SuperAW3000 HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,260.00

€2,260.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€2,260.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

€2,260.00


Please contact Customer Service for Availability

Product Name

TSKgel® SuperAW4000 HPLC Column, phase hydroxylated methacrylate, L × I.D. 15 cm × 6 mm, 6 μm particle size

material

stainless steel hardware

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 19317

parameter

≤0.5 M salt concentration
0-100% polar organic solvent
0.6 mL/min flow rate
40 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

15 cm × 6 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

6 μm

pore size

450 Å

operating pH

2-12

separation technique

size exclusion (SEC)

General description

TSKgel SuperAW4000 columns are packed with rigid, porous, polymer beads which show minimal shrinkage or swelling in different solvent systems. They are used for the separation of polymers that are soluble in polar organic solvents.

Application

Exclusion limit (DMF): Polyethylene oxide 400,000 Da

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service