Skip to Content
Merck
All Photos(1)

Key Documents

818386

TSKgel® SuperQ-5PW (10 µm) HPLC Columns

L x I.D. 7.5 cm × 8 mm, ammonium ion, quaternary phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,050.00

€2,050.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€2,050.00

About This Item

UNSPSC Code:
41115711

€2,050.00


Please contact Customer Service for Availability

Product Name

TSKgel® SuperQ-5PW HPLC Column, phase ammonium ion, quaternary, L × I.D. 7.5 cm × 8 mm, 10 μm particle size, glass

material

glass

Agency

suitable for USP L23

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 18386

parameter

≤0.5 M salt concentration
≤20% organic solvent
≤45 °C temperature
1.2 mL/min flow rate
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

ammonium ion, quaternary phase

particle size

10 μm

operating pH

2-12

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SuperQ-5PW columns are packed with porous hydrophilic polymer beads which are modified with a strong anion exchanger. The resulting product exhibits very high protein capacities. TSKgel SuperQ-5PW is used for the separation and analysis of proteins, oligonucleotides and other biologically active molecules. TSKgel SuperQ-5PW columns have the same backbone chemistry and selectivity as the bulk process scale resins TSKgel SuperQ-5PW (20) and (30) and Toyopearl SuperQ-650.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service