Skip to Content
Merck
All Photos(1)

Key Documents

818278

TSKgel® Super-Phenyl (2.3 µm) HPLC Columns

L x I.D. 10 cm × 4.6 mm, phenyl phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€832.00

€832.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€832.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

€832.00


Please contact Customer Service for Availability

Product Name

TSKgel® Super-Phenyl HPLC Column, phase phenyl, L × I.D. 10 cm × 4.6 mm, 2.3 μm particle size

material

stainless steel hardware

Agency

suitable for USP L11

product line

TSKgel®

manufacturer/tradename

Tosoh Super-Phenyl, 18278

extent of labeling

3% Carbon, multilayer, endcapped

parameter

no limitation salt concentration
0-100% organic solvent
10-50 °C temperature
300 bar max. pressure
4 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

10 cm × 4.6 mm

surface area

100 m2/g

matrix

silica particle platform

matrix active group

phenyl phase

particle size

2.3 μm

pore size

140 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Super-Phenyl columns are packed with monodispersed 2 micron (nominal, mean particle size is 2.3 micron) spherical silica particles covalently bonded with phenyl groups. The small particle size makes the Super series the highest efficiency reversed phase columns in the TSKgel product line. The monodispersed packing generates operational back pressures more typical of larger particles allowing the use of higher flow rates than other 2 micron packings and offers less hydrophobicity than TSKgel Super-Octyl and TSKgel Super-ODS. TSKgel Super-Phenyl is an excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules and food/beverage samples.

Physical form

Shipped in 50% acetonitrile - 50% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service