Skip to Content
Merck
All Photos(1)

Documents

818150

TSKgel® ODS-80Ts (5 µm) HPLC Columns

L x I.D. 15 cm × 2 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® ODS-80Ts HPLC Column, phase C18 (octadecyl), L × I.D. 15 cm × 2 mm, 5 μm particle size

material

stainless steel hardware

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh ODS-80Ts, 18150

extent of labeling

15% Carbon, monolayer, endcapped

parameter

0.22 mL/min flow rate
10-100% organic solvent
10-50 °C temperature
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 2 mm

matrix

silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

80 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel ODS-80Ts is a general purpose column for low MW pharmaceuticals, bases, nucleosides and nucleotides. Common applications include purity checks and peptide mapping. Bonded phase pore size is indicated by the number in the product description, in this case TSKgel ODS-80Ts has 80Ã… nominal pore size. The pore size of the base silica is 100Ã….

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service