Skip to Content
Merck
All Photos(1)

Key Documents

814688

TOYOPEARL® HW-65S Bulk Media

phase hydroxylated methacrylic polymer, bottle of 1 L, 30 μm particle size

Synonym(s):

TOYOPEARL® HW-65 Size Exclusion Media

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€2,710.00

€2,710.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
€2,710.00

About This Item

UNSPSC Code:
41106500

€2,710.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14688

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylic polymer phase

particle size

30 μm

operating pH

2-13

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl HW-65 resin is a hydroxylated methacrylic polymer bead for size exclusion chromatography of proteins between 40,000-5,000,000 Da. It also is the base bead for all Toyopearl "-650" designated products.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service