Skip to Content
Merck
All Photos(1)

Key Documents

80023460

TSKgel® Amide-80 (2 µm) HPLC Columns

L x I.D. 1 cm × 2.0 mm, carbamoyl phase

Synonym(s):

TSKgel® Amide 80 HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€672.00

€672.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€672.00

About This Item

UNSPSC Code:
41115730

€672.00


Please contact Customer Service for Availability

Product Name

TSKgel® Amide 80 Direct Connect Guard Column, phase carbamoyl, L × I.D. 1 cm × 2.0 mm, 2 μm particle size

material

stainless steel

Agency

suitable for USP L68

product line

TSKgel®

manufacturer/tradename

Tosoh 23460

parameter

150 bar max. pressure
50 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2.0 mm

surface area

450 m2/g

matrix

fully porous particle

matrix active group

carbamoyl phase

particle size

2 μm

operating pH range

2.0-7.5

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service