Skip to Content
Merck
All Photos(1)

Documents

796-0658

HPLC Analysis of Proteins on Discovery® BIO PolyMA-SCX (Analyte Set #2)

application for HPLC

Synonym(s):

Proteins, mixture

Sign Into View Organizational & Contract Pricing


About This Item

description

Separated on Discovery® BIO PolyMA-SCX

technique(s)

HPLC: suitable

test parameters

column: Discovery BIO PolyMA-SCX, 5 cm x 4.6 mm, 5 μm particles (59601-U)
mobile phase: [A] 20 mM sodium phosphate, pH 7.0; [B] 20 mM sodium phosphate, 0.5 M NaCl, pH 7.0; 5 to 100% B in 15 min. (linear)
flow rate: 0.5 mL/min
injection: 10 μg trypsin inhibitor, 5 μg conalbumin, myoglobin or 10 μg ribonuclease A, 5 μg cytochrome c, a-chymotrypsinogen A
detector: UV, 280 nm

suitability

application for HPLC

application(s)

life science and biopharma

Analysis Note

The large pore diameter of Discovery BIO PolyMA-SCX and PolyMA-WAX particles allows full access to small peptides and very large proteins and protein aggregates. This Figure shows the power of ion-exchange to resolve proteins with very little difference in molecular weight, an advantage over size-exclusion separations.

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service