Skip to Content
Merck
All Photos(1)

Documents

794-0285

GC Analysis of Terpinen-4-ol and Methyl Mandelate Enantiomers on α-DEX 120, β-DEX 120, and γ-DEX 120

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: 30 m × 0.25 mm I.D., 0.25 μm (α-DEX 24310, β-DEX 24304, γ-DEX 24307)
oven: 130 °C
inj. temp.: 250 °C
detector: FID, 300 °C
carrier gas: helium, 35 cm/sec
injection: 1 μL 100:1 split
sample: 1 mg/mL in methylene chloride

application(s)

chemicals and industrial polymers
flavors and fragrances

Analyte

Description

    Methyl (S)-(+)-mandelate
    Methyl (R)-(−)-mandelate

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service