Skip to Content
Merck
All Photos(1)

Key Documents

74175

Supelco

2-Nitro-1,4-phenylenediamine

analytical standard

Synonym(s):

1,4-Diamino-2-nitrobenzene, 2-Nitro-p-phenylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€52.20

€52.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€52.20

About This Item

Linear Formula:
O2NC6H3(NH2)2
CAS Number:
Molecular Weight:
153.14
Beilstein:
2210195
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

€52.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

135-138 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(N)c(c1)[N+]([O-])=O

InChI

1S/C6H7N3O2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H,7-8H2

InChI key

HVHNMNGARPCGGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>372.2 °F - Pensky-Martens closed cup

Flash Point(C)

> 189 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service