69712
2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-[(hexyl)oxy]-phenol
analytical standard
Synonym(s):
2,4-Diphenyl-6-[2-hydroxy-4-(hexyloxy)phenyl]-1,3,5-triazine, 2-(2-Hydroxy-4-hexyloxyphenyl)-4,6-diphenyl-1,3,5-triazine, 2-[4-(Hexyloxy)-2-hydroxyphenyl]-4,6-diphenyl-1,3,5-triazine, 2,4-Diphenyl-6-[2-hydroxy-4-(hexyloxy)phenyl]-1,3,5-triazine, 2-(2-Hydroxy-4-hexyloxyphenyl)-4,6-diphenyl-1,3,5-triazine, 2-[4-(Hexyloxy)-2-hydroxyphenyl]-4,6-diphenyl-1,3,5-triazine
Select a Size
€60.30
Select a Size
About This Item
€60.30
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
147-151 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
CCCCCCOc1ccc(c(O)c1)-c2nc(nc(n2)-c3ccccc3)-c4ccccc4
InChI
1S/C27H27N3O2/c1-2-3-4-11-18-32-22-16-17-23(24(31)19-22)27-29-25(20-12-7-5-8-13-20)28-26(30-27)21-14-9-6-10-15-21/h5-10,12-17,19,31H,2-4,11,18H2,1H3
InChI key
LEVFXWNQQSSNAC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service