Key Documents
58453
Supelco
Mobile Phase Solvent Debubbler with Bracket
Select a Size
1 EA
€118.00
€118.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€118.00
About This Item
UNSPSC Code:
41115700
€118.00
Please contact Customer Service for Availability
Request a Bulk Order
description
universal design with bracket
General description
Gas bubbles in the mobile phase reduce pump output and cause check valves to malfunction. The mobile phase debubbler removes bubbles from the pump inlet line. Bubbles in the incoming mobile phase rise in the debubbler, displacing an equal volume of liquid from the debubbler reservoir to the pump. When necessary, simply remove the cap and refill the reservoir -- the gas-liquid interface is visible through the housing. Compatible with 1.5 mm I.D. and 3.0 mm I.D. PTFE tubing used with Waters and other popular HPLC pumps. KEL-F, PTFE, and 316 stainless steel construction.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)