Skip to Content
Merck
All Photos(1)

Documents

575608-U

Supelco

Discovery® DSC-Si 96-well solid phase extractio (SPE) Plate

bed wt. 50 mg/well, pk of 1

Synonym(s):

Silica SPE 96-well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

product name

Discovery® DSC-Si SPE 96-well Plate, bed wt. 50 mg/well, pk of 1

material

PE frit (20 μm)
polypropylene hardware

composition

bed wt., 50 mg/well

packaging

pk of 1

technique(s)

solid phase extraction (SPE): suitable

surface area

480 m2/g

matrix

silica gel base material (irregularly shaped, acid washed)

matrix active group

silica bonding

particle size

50 μm

pore size

70 Å pore size

application(s)

food and beverages

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Normal phase
Sample Matrix Compatibility: Organic solvents, oils, and lipids

  • Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques
  • Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions
  • The most polar normal phase sorbent available
  • Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents
  • Available in Büchner Funnel configurations for easy scalability

Application


  • High-Throughput Mass Spectrometry for Biopharma: Utilizes the Discovery DSC-Si SPE 96-well plate for rapid and accurate biomolecule characterization, demonstrating its critical role in high-throughput analytical setups, enhancing productivity in biopharmaceutical analysis (Campuzano et al., 2022).

  • Towards a Point-of-Care Diagnostic Platform for Multiplex Sensing: Highlights the application of Discovery DSC-Si SPE 96-well plates in developing advanced diagnostic platforms, specifically for detecting mild traumatic brain injury biomarkers, showcasing the plate′s versatility and reliability in clinical settings (Jović et al., 2022).

  • Determination of Crizotinib in Plasma by LC-ESI-MS/MS: Demonstrates the use of Discovery DSC-Si SPE 96-well plates in pharmacokinetic studies, providing robust and efficient sample preparation methods for therapeutic drug monitoring and clinical research (Roberts et al., 2014).

  • High-throughput Mass Spectrometric Cytochrome P450 Inhibition Screening: Applies the Discovery DSC-Si SPE 96-well plate in drug metabolism studies, specifically in the screening of cytochrome P450 enzyme inhibitors, which is crucial for predicting drug-drug interactions in drug development (Lim et al., 2013).

  • Development of a High-Throughput SPE/MS Method for Cytochrome P450 Inhibition: Uses the Discovery DSC-Si SPE 96-well plate to enhance throughput in the pharmacological screening of drug candidates, underscoring its importance in the early stages of drug discovery to assess potential toxicities (Lim et al., 2010).

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service