Skip to Content
Merck
All Photos(1)

Documents

57400-U

PHRED Photochemical Reactor

AC/DC input 110 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

General description

PHRED photochemical reactor can be used effectively to improve analyte detection in the presence of interfering matrix components, or when the analytes are present at low concentrations.

Footnote

230 volt CE approved unit available for all the countries. Please contact Supelco Custom Quotations at Supelco_Quotes@sial.com for price and delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service