Key Documents
57316-U
Supelco
SPME Fiber Assembly
greener alternative
for use with manual holder, Nitinol-core (NIT), 24 ga
Select a Size
1 PKG
€539.00
€539.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
€539.00
About This Item
UNSPSC Code:
41115724
NACRES:
NB.21
€539.00
Please contact Customer Service for Availability
Request a Bulk Order
Product Name
SPME Fiber Assembly Bare Fused Silica, needle size 24 ga, for use with manual holder
material
fused silica fiber
plain orange hub
Quality Level
needle size
24 ga
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
solid phase microextraction (SPME): suitable
fiber L
1 cm
application(s)
food and beverages
compatibility
for use with manual holder
greener alternative category
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
Solid phase microextraction (SPME) is a solvent-free sample preparation technique that uses coated fibers to extract analytes of interest from a sample before analysis by GC or HPLC.