Skip to Content
Merck
All Photos(1)

Key Documents

57264-U

Supelco

IonSense® C18 SPE-it Tips

pkg of 96 ea

Synonym(s):

SPME LC TIps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

packaging

pkg of 96 ea

fiber L

1 cm

matrix active group

C18

application(s)

food and beverages

compatibility

for use with pipettor or robotic liquid handling systems

Looking for similar products? Visit Product Comparison Guide

General description

IonSense® C18 SPE-it Tips are solid phase extraction devices configured to permit isolation of analytes from aqueous solutions and then direct analysis without further sample processing.

Legal Information

IonSense is a registered trademark of IonSense, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Esther De La Fuente et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 44(10), 1004-1010 (2009-10-16)
The complex molecular assemblages were analysed in the soil gas phase after applying pine forest wastes (PFW) or sugarbeet vinasses (SBV) for soil-borne crop pests' management. For this purpose, solid-phase micro-extraction (SPME) and gas chromatography (GC) were used coupled with
J Tytgat et al.
International journal of legal medicine, 109(3), 150-154 (1996-01-01)
The most common alkyl nitrites encountered in forensic toxicology are iso-butyl, n-butyl and iso-pentyl(amyl) nitrites. All have become popular as an aphrodisiac, especially among the homosexual population. Alkyl nitrites are a volatile and unstable group of compounds, which hydrolyse in
Jermiah Y Shen et al.
Journal of separation science, 35(19), 2544-2552 (2012-08-22)
The presence of triclosan and triclocarban, two endocrine-disrupting chemicals and antimicrobial agents, and transformation products of triclocarban, 1,3-di(phenyl)urea, 1,3-bis(4-chlorophenyl)urea and 1,3-bis(3,4-dichlorophenyl)urea, in tap water, treated household drinking water, bottled water, and river water samples were investigated using solid-phase micro-extraction coupled
A P Arisseto et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(12), 1832-1839 (2012-08-23)
Selected commercial processed foods available in the Brazilian market (306 samples) were analysed for furan content using a validated gas chromatography-mass spectrometry method preceded by headspace solid phase micro-extraction (HS-SPME-GC/MS). Canned and jarred foods, including vegetable, meat, fruit and sweet
Alfredo Koch et al.
Phytochemistry, 71(17-18), 2190-2198 (2010-10-23)
2-Methoxy-3-isobutylpyrazine (MIBP) contributes a bell pepper aroma to many grape cultivars and has a reported aroma threshold of ∼2 ng L(-1) in water. The purpose of this study was twofold: (1) develop a procedure using headspace solid phase micro-extraction combined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service