57061
Supelclean™ LC-WCX SPE Tube
bed wt. 500 mg, volume 3 mL, pkg of 54 ea
About This Item
Recommended Products
material
polypropylene tube
product line
Supelclean™
composition
bed wt., 500 mg
packaging
pkg of 54 ea
technique(s)
solid phase extraction (SPE): suitable
volume
3 mL
matrix
silica gel
matrix active group
WCX phase
carboxylic acid
diameter
40-45 μm
pore size
60 Å
application(s)
food and beverages
separation technique
ion exchange
ion exchange
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
SPE retention mechanism in this case is based on the electrostatic attraction of charged functional groups of the analyte(s) to oppositely charged functional groups on the sorbent.
Protocols
Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service