Skip to Content
Merck
All Photos(1)

Key Documents

57039

Supelco

Visiprep Manifold Plate (included with standard and DL models unless otherwise noted)

for use with 12-port model (standard and DL), 16 mm test tubes

Synonym(s):

V.M.RACK FOR 16MM TEST TUBES

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€150.00

€150.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 EA
€150.00

About This Item

UNSPSC Code:
12352200

€150.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

technique(s)

solid phase extraction (SPE): suitable

Quality Level

application(s)

food and beverages

compatibility

for use with 12-port model (standard and DL), 16 mm test tubes

Looking for similar products? Visit Product Comparison Guide

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Gardner et al.
Analytical chemistry, 81(2), 557-566 (2008-12-17)
Defensins are highly basic cationic peptides that are important components of the innate and adaptive immune response pathways. In addition, these peptides are involved in CD8+ T cell response to HIV-1, increased pulmonary infection risk among cystic fibrosis patients, upregulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service