Key Documents
56625-U
Supelco
LC-MS Post Column Flow Splitters
Fixed, Split Ratio = 10:1
Synonym(s):
LC-MS Flow Divider
Select a Size
1 EA
€2,030.00
€2,030.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€2,030.00
About This Item
UNSPSC Code:
41121815
NACRES:
SB.52
€2,030.00
Please contact Customer Service for Availability
Request a Bulk Order
General description
LC-MS post column flow splitters with split ratio of 10:1 are ideally used for liquid chromatography with mass spectrometry and capillary chromatography. They generally use two compact fluid resistor elements which are designed as cartridges for easy replacement. They also provide a fixed split ratio with extremely low dead volume. And the split ratio is not affected by changes in solvent viscosity or pressure and is extremely stable and reproducible. Also used for post column applications.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)