Key Documents
55216
Supelco
Frit Removing Tool for Supelco® Analytical Columns
About This Item
UNSPSC Code:
41115700
Application
Removes frits from 2.1 mm I.D. to 4.6 mm I.D. Supelco Discovery and SUPELCOSIL analytical columns.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)